MilliporeSigma
  • Glucose metabolic trapping in mouse arteries: nonradioactive assay of atherosclerotic plaque inflammation applicable to drug discovery.
Glucose metabolic trapping in mouse arteries: nonradioactive assay of atherosclerotic plaque inflammation applicable to drug discovery.
PloS one (2012-12-05)
Richard G Conway, Eyassu Chernet, David C De Rosa, Robert J Benschop, Anne B Need, Emily C Collins, James S Bean, J Michael Kalbfleisch, Mark D Rekhter
ABSTRACT
(18)F-Fluorodeoxyglucose (FDG)-positron emission tomography (PET) imaging of atherosclerosis in the clinic is based on preferential accumulation of radioactive glucose analog in atherosclerotic plaques. FDG-PET is challenging in mouse models due to limited resolution and high cost. We aimed to quantify accumulation of nonradioactive glucose metabolite, FDG-6-phosphate, in the mouse atherosclerotic plaques as a simple alternative to PET imaging. Nonradioactive FDG was injected 30 minutes before euthanasia. Arteries were dissected, and lipids were extracted. The arteries were re-extracted with 50% acetonitrile-50% methanol-0.1% formic acid. A daughter ion of FDG-6-phosphate was quantified using liquid chromatography and mass spectrometry (LC/MS/MS). Thus, both traditional (cholesterol) and novel (FDG-6-phosphate) markers were assayed in the same tissue. FDG-6-phosphate was accumulated in atherosclerotic lesions associated with carotid ligation of the Western diet fed ApoE knockout mice (5.9 times increase compare to unligated carotids, p<0.001). Treatment with the liver X receptor agonist T0901317 significantly (2.1 times, p<0.01) reduced FDG-6-phosphate accumulation 2 weeks after surgery. Anti-atherosclerotic effects were independently confirmed by reduction in lesion size, macrophage number, cholesterol ester accumulation, and macrophage proteolytic activity. Mass spectrometry of FDG-6-phosphate in experimental atherosclerosis is consistent with plaque inflammation and provides potential translational link to the clinical studies utilizing FDG-PET imaging.
Sigma-Aldrich
D-Glucose 6-phosphate solution, ~1 M in H2O (approx. 260 mg per ml)