MilliporeSigma
  • A 2-(2-hydroxyphenyl)-1H-benzimidazole-manganese oxide hybrid as a promising structural model for the tyrosine 161/histidine 190-manganese cluster in photosystem II.
A 2-(2-hydroxyphenyl)-1H-benzimidazole-manganese oxide hybrid as a promising structural model for the tyrosine 161/histidine 190-manganese cluster in photosystem II.
Dalton transactions (Cambridge, England : 2003) (2012-11-28)
Mohammad Mahdi Najafpour, Mahmoud Amouzadeh Tabrizi, Behzad Haghighi, Govindjee
ABSTRACT
In this communication, we report the synthesis, characterization, and electrochemistry of a 2-(2-hydroxyphenyl)-1H-benzimidazole-manganese oxide hybrid. Our results suggest that this compound is a promising model for the manganese cluster together with tyrosine-161 and histidine-190 in photosystem II of plants, algae and cyanobacteria.
Sigma-Aldrich
Benzimidazole, 98%