MilliporeSigma
  • Thromboxane receptors in smooth muscle promote hypertension, vascular remodeling, and sudden death.
Thromboxane receptors in smooth muscle promote hypertension, vascular remodeling, and sudden death.
Hypertension (Dallas, Tex. : 1979) (2012-11-15)
Matthew A Sparks, Natalia A Makhanova, Robert C Griffiths, John N Snouwaert, Beverly H Koller, Thomas M Coffman
ABSTRACT
The prostanoid thromboxane A2 has been implicated to contribute to the pathogenesis of many cardiovascular diseases, including hypertension. To study the role of vascular thromboxane-prostanoid (TP) receptors in blood pressure regulation, we generated mice with cell-specific deletion of TP receptors in smooth muscle using Cre/Loxp technology. We crossed the KISM22α-Cre transgenic mouse line expressing Cre recombinase in smooth muscle cells with a mouse line bearing a conditional allele of the Tbxa2r gene (Tp(flox)). In KISM22α-Cre(+)Tp(flox/flox) (TP-SMKO) mice, TP receptors were efficiently deleted from vascular smooth muscle cells. In TP-SMKOs, acute vasoconstrictor responses to the TP agonist U46619 were attenuated to a similar extent in both the peripheral and renal circulations. Yet, acute vascular responses to angiotensin II were unaffected at baseline and after chronic angiotensin II administration. Infusion of high-dose U46619 caused circulatory collapse and death in a majority of control mice but had negligible hemodynamic effects in TP-SMKOs, which were completely protected from U46619-induced sudden death. Baseline blood pressures were normal in TP-SMKOs. However, the absence of TP receptors in vascular smooth muscle cells was associated with significant attenuation of angiotensin II-induced hypertension and diminished vascular remodeling. This was also associated with reduced urinary thromboxane production after chronic angiotensin II. Thus, TP receptors in vascular smooth muscle cells play a major role in mediating the actions of thromboxane A(2) in TP agonist-induced shock, hypertension, and vascular remodeling of the aorta.
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate