MilliporeSigma
  • Mechanistic and computational studies of exocyclic stereocontrol in the synthesis of bryostatin-like cis-2,6-disubstituted 4-alkylidenetetrahydropyrans by Prins cyclization.
Mechanistic and computational studies of exocyclic stereocontrol in the synthesis of bryostatin-like cis-2,6-disubstituted 4-alkylidenetetrahydropyrans by Prins cyclization.
The Journal of organic chemistry (2012-11-06)
Yasuyuki Ogawa, Phillip P Painter, Dean J Tantillo, Paul A Wender
ABSTRACT
The Prins cyclization of syn-β-hydroxy allylsilanes and aldehydes gives cis-2,6-disubstituted 4-alkylidenetetrahydropyrans as sole products in excellent yields regardless of the aldehyde (R″) or syn-β-hydroxy allylsilane substituent (R') used. By reversing the R″ and R' groups, complementary exocyclic stereocontrol can be achieved. When the anti-β-hydroxy allylsilanes are used, the Prins cyclization gives predominantly cis-2,6-disubstituted 4-alkylidenetetrahydropyrans, now with the opposite olefin geometry in excellent yield. The proposed reaction mechanism and the observed stereoselectivity for these processes are supported by DFT calculations.
Sigma-Aldrich
Bryostatin 1, ≥99%, solid
Pricing and availability is not currently available.