Merck
  • Analytical expressions for the steady-state concentrations of glucose, oxygen and gluconic acid in a composite membrane for closed-loop insulin delivery.
Analytical expressions for the steady-state concentrations of glucose, oxygen and gluconic acid in a composite membrane for closed-loop insulin delivery.
The Journal of membrane biology (2012-11-03)
L Rajendran, L K Bieniasz
ABSTRACT
The mathematical model of Abdekhodaie and Wu (J Membr Sci 335:21-31, 2009) of glucose-responsive composite membranes for closed-loop insulin delivery is discussed. The glucose composite membrane contains nanoparticles of an anionic polymer, glucose oxidase and catalase embedded in a hydrophobic polymer. The model involves the system of nonlinear steady-state reaction-diffusion equations. Analytical expressions for the concentration of glucose, oxygen and gluconic acid are derived from these equations using the Adomian decomposition method. A comparison of the analytical approximation and numerical simulation is also presented. An agreement between analytical expressions and numerical results is observed.
Product Description
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Iron(II) D-gluconate dihydrate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium D-gluconate, SAJ first grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Pricing and availability is not currently available.
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.