MilliporeSigma
  • Synergism and rules from combination of Baicalin, Jasminoidin and Desoxycholic acid in refined Qing Kai Ling for treat ischemic stroke mice model.
Synergism and rules from combination of Baicalin, Jasminoidin and Desoxycholic acid in refined Qing Kai Ling for treat ischemic stroke mice model.
PloS one (2012-10-11)
Jian Li, Run-guo Wu, Fan-yun Meng, Zhong Wang, Chang-ming Wang, Yong-yan Wang, Zhan-jun Zhang
ABSTRACT
Refined Qing-Kai-Ling (QKL), a modified Chinese medicine, consists of three main ingredients (Baicalin, Jasminoidin and Desoxycholic acid), plays a synergistic effect on the treatment of the acute stage of ischemic stroke. However, the rules of the combination and synergism are still unknown. Based on the ischemic stroke mice model, all different kinds of combination of Baicalin, Jasminoidin, and Desoxycholic acid were investigated by the methods of neurological examination, microarray, and genomics analysis. As a result, it confirmed that the combination of three drugs offered a better therapeutical effect on ischemic stroke than monotherapy of each drug. Additionally, we used Ingenuity pathway Analysis (IPA) and principal component analysis (PCA) to extract the dominant information of expression changes in 373 ischemia-related genes. The results suggested that 5 principal components (PC1-5) could account for more than 95% energy in the gene data. Moreover, 3 clusters (PC1, PC2+PC5, and PC3+PC4) were addressed with cluster analysis. Furthermore, we matched PCs on the drug-target networks, the findings demonstrated that Baicalin related with PC1 that played the leading role in the combination; Jasminoidin related with PC2+PC5 that played a compensatory role; while Desoxycholic acid had the least performance alone which could relate with PC3+PC4 that played a compatible role. These manifestations were accorded with the principle of herbal formulae of Traditional Chinese Medicine (TCM), emperor-minister-adjuvant-courier. In conclusion, we firstly provided scientific evidence to the classic theory of TCM formulae, an initiating holistic viewpoint of combination therapy of TCM. This study also illustrated that PCA might be an applicable method to analyze the complicated data of drug combination.
Sigma-Aldrich
Geniposide, ≥98% (HPLC)