MilliporeSigma
Preparation of micelle-forming polymer-drug conjugates.
Bioconjugate chemistry (1992-07-01)
M Yokoyama, G S Kwon, T Okano, Y Sakurai, T Seto, K Kataoka
ABSTRACT
Adriamycin, a hydrophobic anticancer drug, was conjugated with poly(ethylene oxide)-poly(aspartic acid) block copolymers composed of various lengths of each block copolymer segment ranging from 1000 to 12,000 in molecular weight and from 10 to 80 units, respectively. Conjugation was achieved without precipitation by adjusting the ratio of adriamycin to aspartic acid residues of the block copolymer and the quantity of DMF used for the reaction. Thus obtained conjugates showed high water solubility irrespective of a large amount of the conjugated adriamycin. Furthermore, these conjugates were found to form micellar structures with a hydrophobic inner core and a hydrophilic outer shell. This micellar architecture may be utilized for effective drug targeting.
Sigma-Aldrich
Methoxypolyethylene glycol amine, 750
Pricing and availability is not currently available.