MilliporeSigma
  • Sprayed nanostructured TiO2 films for efficient photocatalytic degradation of textile azo dye.
Sprayed nanostructured TiO2 films for efficient photocatalytic degradation of textile azo dye.
Journal of photochemistry and photobiology. B, Biology (2012-09-29)
Irina Stambolova, Capital Em Cyrillicaria Shipochka, Vladimir Blaskov, Alexandrе Loukanov, Sasho Vassilev
ABSTRACT
Spray pyrolysis procedure for preparation of nanostructured TiO(2) films with higher photocatalytic effectiveness and longer exploitation life is presented in this study. Thin films of active nanocrystalline TiO(2) were obtained from titanium isopropoxide, stabilized with acetyl acetone and characterized by X-ray photoelectron spectroscopy (XPS), scanning electron microscopy (SEM) and X-ray diffraction (XRD). The activity of sprayed nanostructured TiO(2) is tested for photocatalytic degradation of Reactive Black 5 dye with concentrations up to 80 ppm. Interesting result of the work is the reduction of toxicity after photocatalytic treatment of RB5 with TiO(2), which was confirmed by the lower percentage of mortality of Artemia salina. It was proved that the film thickness, conditions of post deposition treatment and the type of the substrate affected significantly the photocatalytic reaction. Taking into account that the parameters are interdependent, it is necessary to optimize the preparation conditions in order to synthesize photocatalytic active films.
Sigma-Aldrich
Reactive Black 5, Dye content ≥50 %
Pricing and availability is not currently available.