MilliporeSigma
  • Accumulation of squalene is associated with the clustering of lipid droplets.
Accumulation of squalene is associated with the clustering of lipid droplets.
The FEBS journal (2012-09-28)
Minh T Ta, Tamar S Kapterian, Weihua Fei, Ximing Du, Andrew J Brown, Ian W Dawes, Hongyuan Yang
ABSTRACT
The isoprenoid squalene is an important precursor for the biosynthesis of sterols. The cellular storage of squalene and its impact on membrane properties have been the subject of recent investigations. In a screen for abnormal lipid droplet morphology and distribution in the yeast Saccharomyces cerevisiae, we found significant lipid droplet clustering (arbitrarily defined as an aggregation of six or more lipid droplets) in a number of mutants (e.g. erg1) that are defective in sterol biosynthesis. Interestingly, these mutants are also characterized by accumulation of large amounts of squalene. Reducing the level of squalene in these mutants restored normal lipid droplet distribution. Moreover, inhibition of squalene monooxygenase in two mammalian cell lines (CHO-K1 and 3T3-L1) by terbinafine also resulted in lipid droplet clustering. These results indicate that the level of squalene may affect the growth and distribution of lipid droplets.
Sigma-Aldrich
Squalene, ≥98%, liquid
Supelco
Squalene, analytical standard