MilliporeSigma
  • Is basal ultrasensitive measurement of calcitonin capable of substituting for the pentagastrin-stimulation test?
Is basal ultrasensitive measurement of calcitonin capable of substituting for the pentagastrin-stimulation test?
Clinical endocrinology (2012-08-24)
Géraldine Pina, Séverine Dubois, Arnaud Murat, Nicole Berger, Patricia Niccoli, Jean-Louis Peix, Régis Cohen, Claudine Guillausseau, Anne Charrie, Olivier Chabre, Catherine Cornu, Françoise Borson-Chazot, Vincent Rohmer
ABSTRACT
To evaluate a second-generation assay for basal serum calcitonin (CT) measurements compared with the pentagastrin-stimulation test for the diagnosis of inherited medullary thyroid carcinoma (MTC) and the follow-up of patients with MTC after surgery. Recent American Thyroid Association recommendations suggest the use of basal CT alone to diagnose and assess follow-up of MTC as the pentagastrin (Pg) test is unavailable in many countries. Multicentric prospective study. A total of 162 patients with basal CT <10 ng/l were included: 54 asymptomatic patients harboured noncysteine 'rearranged during transfection' (RET) proto-oncogene mutations and 108 patients had entered follow-up of MTC after surgery. All patients underwent basal and Pg-stimulated CT measurements using a second-generation assay with 5-ng/l functional sensitivity. Ninety-five per cent of patients with basal CT ≥ 5 ng/l and 25% of patients with basal CT <5 ng/l had a positive Pg-stimulation test (Pg CT >10 ng/l). Compared with the reference Pg test, basal CT ≥ 5 ng/l had 99% specificity, a 95%-positive predictive value but only 35% sensitivity (P < 0.0001). Overall, there were 31% less false-negative results using a 5-ng/l threshold for basal CT instead of the previously used 10-ng/l threshold. The ultrasensitive CT assay reduces the false-negative rate of basal CT measurements when diagnosing familial MTC and in postoperative follow-up compared with previously used assays. However, its sensitivity to detect C-cell disease remains lower than that of the Pg-stimulation test.
Sigma-Aldrich
Pentagastrin, ≥95% (HPLC), powder