MilliporeSigma
  • Characterization of cobalt(III) hydroxamic acid complexes based on a tris(2-pyridylmethyl)amine scaffold: reactivity toward cysteine methyl ester.
Characterization of cobalt(III) hydroxamic acid complexes based on a tris(2-pyridylmethyl)amine scaffold: reactivity toward cysteine methyl ester.
Inorganic chemistry (2012-08-22)
Mickael Alimi, Anas Allam, Mohamed Selkti, Alain Tomas, Pascal Roussel, Erwan Galardon, Isabelle Artaud
ABSTRACT
Six Co(III) complexes based on unsubstituted or substituted TPA ligands (where TPA is tris(2-pyridylmethyl)amine) and acetohydroxamic acid (A), N-methyl-acetohydroxamic acid (B), or N-hydroxy-pyridinone (C) were prepared and characterized by mass spectrometry, elemental analysis, and electrochemistry: [Co(III)(TPA)(A-2H)](Cl) (1a), [Co(III)((4-Cl(2))TPA)(A-2H)](Cl) (2a), [Co(III)((6-Piva)TPA)(A-2H)](Cl) (3a), [Co(III)((4-Piva)TPA)(A-2H)](Cl) (4a) and [Co(III)(TPA)(B-H)](Cl)(2) (1b), and [Co(III)(TPA)(C-H)](Cl)(2) (1c). Complexes 1a-c and 3a were analyzed by (1)H NMR, using 2D ((1)H, (1)H) COSY and 2D ((1)H, (13)C) HMBC and HSQC, and shown to exist as a mixture of two geometric isomers based on whether the hydroxamic oxygen was trans to a pyridine nitrogen or to the tertiary amine nitrogen. Complex 3a exists as a single isomer that was crystallized. Its crystal structure revealed the presence of an H-bond between the pivaloylamide and the hydroximate oxygen. Complexes 1a, 2a, and 4a are irreversibly reduced beyond -900 mV versus SCE, while complexes 1b and 1c are reduced at less negative values of -330 and -190 mV, respectively. The H-bond in 3a increased the redox potential up to -720 mV. Reaction of complex 1a with L-cysteine methyl ester CysOMe was monitored by (1)H NMR and UV-vis at 2 mM and 0.2 mM in an aqueous buffered solution at pH 7.5. Complex 1a was successively converted into an intermediate [Co(III)(TPA)(CysOMe-H)](2+), 1d, by exchange of the hydroximate with the cysteinate ligand, and further into Co(III)(CysOMe-H)(3), 5. An authentic sample of 1d was prepared and thoroughly characterized. A detailed (1)H NMR analysis showed there was only one isomer, in which the thiolate was trans to the tertiary amine nitrogen.
Sigma-Aldrich
L-Cysteine methyl ester hydrochloride, 98%
Pricing and availability is not currently available.