MilliporeSigma
  • Induction of hexanol dehydrogenase in Geotrichum spp. by the addition of hexanol.
Induction of hexanol dehydrogenase in Geotrichum spp. by the addition of hexanol.
Applied microbiology and biotechnology (2012-07-24)
Jinhua Zhang, Junling Shi, Hongliang Lv, Yanlin Liu
ABSTRACT
Excessive hexanol content distorts the flavor of foods and is harmful to human health. Previously, two strains of fungi were found capable of producing hexanol-degrading enzymes. The current study identified these strains as Galactomyces geotrichum according to the gene sequence of the 26 S rDNA D1/D2 region (strain S12) and genus Geotrichum according to the gene sequence of ITS region (strain S13). Parallel analysis of extracellular and intracellular enzyme activities showed that the enzymes mainly accumulated intracellularly. Native polyacrylamide gel electrophoresis with reactive dyes showed the enzymes were alcohol dehydrogenases induced by the addition of hexanol. Hexanol was catalyzed into hexanoic acid and hexanal by strain S12 and into hexanoic acid by strain S13. The optimum conditions for the induction of enzymes were determined to be 6-9 h in the presence of 0.7 g/l hexanol. The identification of two strains capable of enzymatically degrading hexanol and optimum conditions for their induction will facilitate their use in industrial applications.
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Supelco
Hexanoic acid, analytical standard
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium hexanoate, 99-100%
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Hexanal, ≥97%, FCC, FG
Sigma-Aldrich
Hexanal, 98%