MilliporeSigma
  • Synthesis and antiangiogenic activity of novel gambogic acid derivatives.
Synthesis and antiangiogenic activity of novel gambogic acid derivatives.
Molecules (Basel, Switzerland) (2012-05-29)
Tao Chen, Rong-Hong Zhang, Shi-Chao He, Qin-Yuan Xu, Liang Ma, Guang-Cheng Wang, Neng Qiu, Fei Peng, Jin-Ying Chen, Jing-Xiang Qiu, Ai-Hua Peng, Li-Juan Chen
ABSTRACT
Gambogic acid (GA) is in a phase II clinical trial as an antitumor and antiangiogenesis agent. In this study, 36 GA derivatives were synthesized and screened in a zebrafish model to evaluate their antiangiogenic activity and toxicity. Derivatives 4, 32, 35, 36 effectively suppressed the formation of newly grown blood vessels and showed lower toxicities than GA as evaluated by zebrafish heart rates and mortalities. They also exhibited more potent migration and HUVEC tube formation inhibiting activities than GA. Among them, 36 was the most potent one, suggesting that it may serve as a potential new antiangiogenesis candidate with low toxicity. Additionally, 36 showed comparable antiproliferative activity to HUVECs and five tumor cell lines but low cytotoxicity to LO2 cells.