MilliporeSigma
  • Polysulfone functionalized with phosphonated poly(pentafluorostyrene) grafts for potential fuel cell applications.
Polysulfone functionalized with phosphonated poly(pentafluorostyrene) grafts for potential fuel cell applications.
Macromolecular rapid communications (2012-05-25)
Ivaylo Dimitrov, Shogo Takamuku, Katja Jankova, Patric Jannasch, Søren Hvilsted
ABSTRACT
A multi-step synthetic strategy to polysulfone (PSU) grafted with phosphonated poly(pentafluorostyrene) (PFS) is developed. It involves controlled radical polymerization resulting in alkyne-end functional PFS. The next step is the modification of PSU with a number of azide side groups. The grafting of PFS onto PSU backbone is performed via the "click"-chemistry approach. In a final step, the PFS-grafts are subjected to the post phosphonation. The copolymers are evaluated as membranes for potential fuel cell applications through thermal analyses, water uptake, and conductivity measurements. The proposed synthetic route opens the possibility to tune copolymers' hydrophilic-hydrophobic balance to obtain membranes with an optimal balance between proton conductivity and mechanical properties.
Sigma-Aldrich
Polystyrene, average Mw 35,000
Pricing and availability is not currently available.
Sigma-Aldrich
Polystyrene, average Mw ~280,000 by GPC
Pricing and availability is not currently available.
Sigma-Aldrich
Polystyrene, average Mw ~400,000 by GPC
Pricing and availability is not currently available.
Sigma-Aldrich
Polystyrene, average Mw ~192,000
Pricing and availability is not currently available.
Sigma-Aldrich
Polystyrene, average Mw ~350,000, average Mn ~170,000
Pricing and availability is not currently available.
Sigma-Aldrich
Polysulfone, average Mn ~22,000 by MO, beads
Pricing and availability is not currently available.
Sigma-Aldrich
Polysulfone, average Mw ~35,000 by LS, average Mn ~16,000 by MO, pellets (Transparent)
Pricing and availability is not currently available.