MilliporeSigma
  • Diastereo- and enantioselective [3 + 2] cycloaddition reaction of Morita-Baylis-Hillman carbonates of isatins with N-phenylmaleimide catalyzed by Me-DuPhos.
Diastereo- and enantioselective [3 + 2] cycloaddition reaction of Morita-Baylis-Hillman carbonates of isatins with N-phenylmaleimide catalyzed by Me-DuPhos.
The Journal of organic chemistry (2012-04-05)
Yan Wang, Li Liu, Tao Zhang, Neng-Jun Zhong, Dong Wang, Yong-Jun Chen
ABSTRACT
A Me-DuPhos-catalyzed efficient asymmetric [3 + 2] cycloaddition reaction between Morita-Baylis-Hillman carbonates of isatins and N-phenylmaleimide has been developed. This reaction constructs three chiral centers in one step to afford spirocyclopentaneoxindoles in good yields (up to 84%) with excellent diastereo- and enantioselectivies (up to 99% ee).
Sigma-Aldrich
N-Phenylmaleimide, 97%