MilliporeSigma
Patulin: Mechanism of genotoxicity.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-03-20)
Nina Glaser, Helga Stopper
ABSTRACT
Patulin is a frequently found food contaminant mainly produced by the fungi Aspergillus and Penicillium. Patulin is suspected to be clastogenic, mutagenic, teratogenic and in higher concentrations cytotoxic. Here, we investigate the mechanism of the patulin-induced genotoxicity. Chromosomal damage was detected as micronucleus and nucleoplasmic bridge formation. Due to the activity of patulin on SH-groups, glutathione is a major compound in the cellular defense against patulin and the depletion of glutathione level with buthionine sulfoximine led to a strong increase in the genoxicity of patulin. A modified version of the alkaline comet assay was carried out to show the cross-linking properties of patulin. As a mechanistic hypothesis, we suspect patulin to cause structural DNA damage by cross-linking, yielding nucleoplasmic bridges and as a later consequence, micronucleus formation. The structural DNA damage may also lead to cell cycle delays, the consequence of which may be the observed centrosome amplification and formation of multipolar mitotic spindles.
Product Description
Supelco
Patulin solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Patulin, ≥98.0% (HPLC)