MilliporeSigma
  • Impact of the CATIE trial on antipsychotic prescribing patterns at a state psychiatric facility.
Impact of the CATIE trial on antipsychotic prescribing patterns at a state psychiatric facility.
Schizophrenia research (2012-03-01)
Angela M Koranek, Tawny L Smith, Lisa M Mican, Karen L Rascati
ABSTRACT
Results from the Clinical Antipsychotic Trials of Intervention Effectiveness (CATIE) indicate that, with the exception of olanzapine, no substantial overall differences were identified between second generation antipsychotics (SGAs) and the first generation antipsychotic (FGA) perphenazine. This study evaluated the effect of CATIE on antipsychotic prescribing. A retrospective review of 1807 adults with schizophrenia was conducted and relative quarterly percentages of FGA versus SGA prescriptions were calculated. Time series analysis did not identify significant differences in rates of FGA prescriptions. Critiques of the methods used in CATIE may have mitigated its potential impact on antipsychotic prescribing despite cost-effectiveness of perphenazine treatment.
Sigma-Aldrich
Perphenazine
Pricing and availability is not currently available.