MilliporeSigma
  • DNA binding and antiproliferative activity toward human carcinoma cells of copper(II) and zinc(II) complexes of a 2,5-diphenyl[1,3,4]oxadiazole derivative.
DNA binding and antiproliferative activity toward human carcinoma cells of copper(II) and zinc(II) complexes of a 2,5-diphenyl[1,3,4]oxadiazole derivative.
Dalton transactions (Cambridge, England : 2003) (2012-02-16)
Alessio Terenzi, Mirco Fanelli, Gianluca Ambrosi, Stefano Amatori, Vieri Fusi, Luca Giorgi, Vincenzo Turco Liveri, Giampaolo Barone
ABSTRACT
The interaction of calf thymus DNA with [CuL(ClO(4))]ClO(4)·H(2)O (1) and [ZnLBr]Br·H(2)O (2) (L = 9,12,15,18,27,28-hexaaza-29-oxatetracyclo[24.2.1.0(2,7).0(20,25)]enneicosa-2,4,6,20,22,24,26,28(1)-octaene) dicationic complexes in aqueous solution at neutral pH, was investigated by variable-temperature UV-vis absorption, circular dichroism and fluorescence spectroscopy. The values of the DNA-binding constants of these complexes, determined by competitive binding spectrofluorimetric titrations of ethidium bromide (EB)-DNA solutions, are (6.7 ± 0.5) × 10(6) M(-1) for CuL(2+) and (4.7 ± 0.5) × 10(5) M(-1) for ZnL(2+). These data together with a through analysis of the spectroscopic behaviour consistently suggest that both compounds are effective DNA binders. Interestingly, the DNA-binding strength of these complexes has been found to be correlated to their in vitro cytotoxic activity toward human breast carcinoma cells, although the complex with lower DNA-binding affinity is more active. In fact, biological studies showed that when the compounds are delivered through the cell membrane by a lipidic carrier, the cell survival is sensibly reduced, up to 58% with 1 and to 31% with 2.
Sigma-Aldrich
2,5-Diphenyloxazole, suitable for liquid scintillation spectrometry
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,5-Diphenyloxazole, 99%, suitable for scintillation