MilliporeSigma
Antioxidant properties of ethyl vanillin in vitro and in vivo.
Bioscience, biotechnology, and biochemistry (2011-12-08)
Akihiro Tai, Takeshi Sawano, Futoshi Yazama
ABSTRACT
We systematically evaluated the antioxidant activity of ethyl vanillin, a vanillin analog, as compared with the activities of vanillin and other vanillin analogs using multiple assay systems. Ethyl vanillin and vanillin exerted stronger antioxidant effects than did vanillyl alcohol or vanillic acid in the oxygen radical absorbance capacity (ORAC) assay, although the antioxidant activities of vanillyl alcohol and vanillic acid were clearly superior to those of ethyl vanillin and vanillin in the three model radical assays. The antioxidant activity of ethyl vanillin was much stronger than that of vanillin in the oxidative hemolysis inhibition assay, but was the same as that of vanillin in the ORAC assay. Oral administration of ethyl vanillin to mice increased the concentration of ethyl vanillic acid, and effectively raised antioxidant activity in the plasma as compared to the effect of vanillin. These data suggest that the antioxidant activity of ethyl vanillin might be more beneficial than has been thought in daily health practice.
Sigma-Aldrich
Ethyl vanillin, ≥98%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
3-Ethoxy-4-hydroxybenzaldehyde, ReagentPlus®, 99%
Pricing and availability is not currently available.