MilliporeSigma
  • Inhibition of fatty acid synthase by amentoflavone reduces coxsackievirus B3 replication.
Inhibition of fatty acid synthase by amentoflavone reduces coxsackievirus B3 replication.
Archives of virology (2011-11-15)
Steffi Wilsky, Katharina Sobotta, Nadine Wiesener, Johanna Pilas, Nadine Althof, Thomas Munder, Peter Wutzler, Andreas Henke
ABSTRACT
Coxsackievirus B3 (CVB3) is a human pathogen that causes acute and chronic infections, but an antiviral drug to treat these diseases has not yet been developed for clinical use. Several intracellular pathways are altered to assist viral transcription, RNA replication, and progeny release. Among these, fatty acid synthase (FAS) expression is increased. In order to test the potential of FAS inhibition as an anti-CVB3 strategy, several experiments were performed, including studies on the correlation of CVB3 replication and FAS expression in human Raji cells and an analysis of the time and dose dependence of the antiviral effect of FAS inhibition due to treatment with amentoflavone. The results demonstrate that CVB3 infection induces an up-regulation of FAS expression already at 1 h postinfection (p.i.). Incubation with increasing concentrations of amentoflavone inhibited CVB3 replication significantly up to 8 h p.i. In addition, suppression of p38 MAP kinase activity by treatment with SB239063 decreased FAS expression as well as viral replication. These data provide evidence that FAS inhibition via amentoflavone administration might present a target for anti-CVB3 therapy.
Product Description
Supelco
Amentoflavone, analytical standard
Sigma-Aldrich
Amentoflavone, ≥98.0% (HPLC)