MilliporeSigma
  • Simultaneous determination of norfloxacin and lomefloxacin in milk by first derivative synchronous fluorescence spectrometry using Al (III) as an enhancer.
Simultaneous determination of norfloxacin and lomefloxacin in milk by first derivative synchronous fluorescence spectrometry using Al (III) as an enhancer.
Analytica chimica acta (2011-10-27)
Yan-Ni Yi, Gui-Rong Li, Yong-Sheng Wang, Yu-Zhen Zhou, Hui-Min Zhu
ABSTRACT
A novel method for the simultaneous determination of norfloxacin (NFLX) and lomefloxacin (LFLX) in milk samples was developed by using first derivative synchronous fluorimetry. The synchronous fluorescence (Δλ=160 nm) spectra and first derivative synchronous fluorescence spectra of NFLX, LFLX and their mixture were studied. The zero-crossing method was utilized to measure the first derivative value of the derivative spectrum. The zero-crossing points were located at 275.0 nm for NFLX and at 283.8 nm for LFLX, in first derivative synchronous fluorescence spectra. Therefore, 283.8 nm and 275.0 nm were selected for the determination of NFLX and LFLX. The first derivative values varied linearly with the concentrations in the range of 1.68×10(-8)-5.64×10(-6) mol L(-1) for NFLX and 1.89×10(-8)-6.19×10(-6) mol L(-1) for LFLX. The detection limits were 5.03×10(-9) mol L(-1) for NFLX and 7.58×10(-9) mol L(-1) for LFLX. The proposed method is reliable, selective and sensitive, and has been used successfully in the simultaneous determination of NFLX and LFLX in milk samples, whose results were in good agreement with those obtained by HPLC.
Sigma-Aldrich
Lomefloxacin hydrochloride