MilliporeSigma
  • Nitrendipine nanocrystals: its preparation, characterization, and in vitro-in vivo evaluation.
Nitrendipine nanocrystals: its preparation, characterization, and in vitro-in vivo evaluation.
AAPS PharmSciTech (2011-09-06)
Peng Quan, Dengning Xia, Hongze Piao, Hongyu Piao, Kai Shi, Yinnong Jia, Fude Cui
ABSTRACT
The present investigation was undertaken with the objective of developing a solid formulation containing nitrendipine nanocrystals for oral delivery. Nitrendipine nanocrystals were prepared using a tandem precipitation-homogenization process. Then, spray drying, a cost-effective method very popular in industrial situations, was employed to convert the nanocrystals into a solid form. The parameters of the preparation process were investigated and optimized. The optimal process was as follows: firstly, nitrendipine/acetone solution (100 mg/ml) was added to a polyvinyl alcohol solution (1 mg/ml) at 10°C, then the pre-suspension was homogenized for 20 cycles at 1,000 bar. Both differential scanning calorimetry and X-ray diffraction analysis indicated that nitrendipine was present in crystalline form. The in vitro dissolution rate of the nanocrystals was significantly increased compared with the physical mixture and commercial tablet. The in vivo testing demonstrated that the C(max) of the nanocrystals was approximately 15-fold and 10-fold greater than that of physical mixture and commercial tablet, respectively. In addition, the AUC(0→24) of the nanocrystals was approximately 41-fold and 10-fold greater than that of physical mixture and commercial tablet, respectively.
Sigma-Aldrich
Nitrendipine, >95%, powder
Pricing and availability is not currently available.