MilliporeSigma
  • Microbial utilization and selectivity of pectin fractions with various structures.
Microbial utilization and selectivity of pectin fractions with various structures.
Applied and environmental microbiology (2011-07-05)
Chatchaya Onumpai, Sofia Kolida, Estelle Bonnin, Robert A Rastall
ABSTRACT
To evaluate the fermentation properties of oligosaccharides derived from pectins and their parent polysaccharides, a 5-ml-working-volume, pH- and temperature-controlled fermentor was tested. Six pectic oligosaccharides representing specific substructures found within pectins were prepared. These consisted of oligogalacturonides (average degrees of polymerization [DP] of 5 and 9), methylated oligogalacturonides (average DP of 5), oligorhamnogalacturonides (average DP of 10 as a disaccharide unit of galacturonic acid and rhamnose), oligogalactosides (average DP of 5), and oligoarabinosides (average DP of 6). The influence of these carbohydrates on the human fecal microbiota was evaluated. Use of neutral sugar fractions resulted in an increase in Bifidobacterium populations and gave higher organic acid yields. The Bacteroides-Prevotella group significantly increased on all oligosaccharides except oligogalacturonides with an average DP of 5. The most selective substrates for bifidobacteria were arabinan, galactan, oligoarabinosides, and oligogalactosides.
Sigma-Aldrich
D-Galacturonic acid sodium salt, ≥95.0% (T)