MilliporeSigma
  • Activated carbons from sisal waste by chemical activation with K₂CO₃: kinetics of paracetamol and ibuprofen removal from aqueous solution.
Activated carbons from sisal waste by chemical activation with K₂CO₃: kinetics of paracetamol and ibuprofen removal from aqueous solution.
Bioresource technology (2011-07-05)
Ana S Mestre, Ana S Bexiga, Margarida Proença, Marta Andrade, Moisés L Pinto, Inês Matos, Isabel M Fonseca, Ana P Carvalho
ABSTRACT
Sisal waste was used as precursor to prepare carbons by chemical activation. The influence of the K(2)CO(3) amount and activation temperature on the materials textural properties were studied through N(2) and CO(2) adsorption assays. As the severity of the treatment increases there is a development of supermicropores, and the micropore size distribution changes from mono to bimodal. A carbon with an apparent surface area of 1,038 m(2)g(-1) and pore volume of 0.49 cm(3)g(-1) was obtained. TPD results showed the incidence in acidic type groups although the pH(PZC) reveals an almost neutral character of the surface. Adsorption kinetic data of ibuprofen and paracetamol show that the processes obey to a pseudo-second order kinetic equation. Regarding the removal efficiency the prepared samples attained values comparable to a commercial carbon (>65%), revealing that chemical activation of sisal wastes with K(2)CO(3) allows obtaining samples suitable for pharmaceutical compounds removal from liquid phase.
Sigma-Aldrich
Potassium carbonate, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, −325 mesh, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Potassium carbonate, puriss., meets analytical specification of Ph. Helv., anhydrous, granulated, 99-101% (calc. to the dried substance)
Sigma-Aldrich
Potassium carbonate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh