MilliporeSigma
  • A key enzyme in the biogenesis of lysosomes is a protease that regulates cholesterol metabolism.
A key enzyme in the biogenesis of lysosomes is a protease that regulates cholesterol metabolism.
Science (New York, N.Y.) (2011-07-02)
Katrin Marschner, Katrin Kollmann, Michaela Schweizer, Thomas Braulke, Sandra Pohl
ABSTRACT
Mucolipidosis II is a severe lysosomal storage disorder caused by defects in the α and β subunits of the hexameric N-acetylglucosamine-1-phosphotransferase complex essential for the formation of the mannose 6-phosphate targeting signal on lysosomal enzymes. Cleavage of the membrane-bound α/β-subunit precursor by an unknown protease is required for catalytic activity. Here we found that the α/β-subunit precursor is cleaved by the site-1 protease (S1P) that activates sterol regulatory element-binding proteins in response to cholesterol deprivation. S1P-deficient cells failed to activate the α/β-subunit precursor and exhibited a mucolipidosis II-like phenotype. Thus, S1P functions in the biogenesis of lysosomes, and lipid-independent phenotypes of S1P deficiency may be caused by lysosomal dysfunction.
Sigma-Aldrich
D-Mannose 6-phosphate sodium salt, ≥98% (HPLC)
Sigma-Aldrich
D-Mannose 6-phosphate disodium salt hydrate, ≥97.0% dry basis (enzymatic)