MilliporeSigma
  • Differentiation of Cuscuta chinensis and Cuscuta australis by HPLC-DAD-MS analysis and HPLC-UV quantitation.
Differentiation of Cuscuta chinensis and Cuscuta australis by HPLC-DAD-MS analysis and HPLC-UV quantitation.
Planta medica (2011-06-16)
Xianghui He, Wenzhi Yang, Min Ye, Qing Wang, Dean Guo
ABSTRACT
Cuscuta chinensis and Cuscuta australis, the two botanical sources of the Chinese herbal medicine Tu-Si-Zi, were distinguished from each other based on qualitative and quantitative chemical analysis. By HPLC‑DAD‑MS, a total of 36 compounds were characterized from these two Cuscuta species, including 14 flavonoids, 17 quinic acid derivatives, and 5 lignans. In addition, HPLC‑UV was applied to determine seven major compounds (6 flavonoids plus chlorogenic acid) in 27 batches of Tu-Si-Zi. The results revealed that the amounts of the three classes of compounds varied significantly between the species. C. australis contained more flavonoids but less quinic acid derivatives and lignans than C. chinensis. Particularly, the amounts of kaempferol and astragalin in C. australis were remarkably higher than in C. chinensis. This finding could be valuable for the quality control of Tu-Si-Zi.
Sigma-Aldrich
Kaempferol 3-β-D-glucopyranoside, ≥97.0% (HPLC)
Pricing and availability is not currently available.