MilliporeSigma
  • Effects of creatine and its analog, β-guanidinopropionic acid, on the differentiation of and nucleoli in myoblasts.
Effects of creatine and its analog, β-guanidinopropionic acid, on the differentiation of and nucleoli in myoblasts.
Bioscience, biotechnology, and biochemistry (2011-06-15)
Yoshinobu Ohira, Yoshikazu Matsuoka, Fuminori Kawano, Akihiko Ogura, Yoko Higo, Takashi Ohira, Masahiro Terada, Yoshihiko Oke, Naoya Nakai
ABSTRACT
The effects of supplementation with creatine (Cr) and its analog, β-guanidinopropionic acid (β-GPA), on the differentiation of myoblasts and the numbers of nucleoli were studied in C2C12 cells. The cells were cultured in differentiation medium for 4 d. Then Cr (1 mM) or β-GPA (1 mM) was added to the cells, and the mixture was cultured for an additional 2 d. Although the number of myotubes was not different among the groups, myotube diameters and nuclear numbers in myotubes were increased by Cr and β-GPA treatment respectively. The expression of differentiation marker proteins, myogenin, and the myosine heavy chain, was increased in the β-GPA group. Supplementation with β-GPA also increased the percentage of p21 (inhibitor for cell cycle progression)-positive myoblasts. Supplementation with Cr inhibited the decrease in nucleoli numbers, whereas β-GPA increased nucleolar sizes in the myotubes. These results suggest that β-GPA supplementation stimulated the differentiation of myoblasts into multi-nucleated myotubes through induction of p21 expression.
Sigma-Aldrich
3-Guanidinopropionic acid