MilliporeSigma
  • Chemical constituents from Lobelia chinensis and their anti-virus and anti-inflammatory bioactivities.
Chemical constituents from Lobelia chinensis and their anti-virus and anti-inflammatory bioactivities.
Archives of pharmacal research (2011-06-10)
Ping-Chung Kuo, Tsong-Long Hwang, Ying-Ting Lin, Yuh-Chi Kuo, Yann-Lii Leu
ABSTRACT
In total, forty six compounds, including the novel compound lobechine (1), were characterized from the methanol extracts of Lobelia chinensis. The chemical structures of known metabolites were identified by comparing their spectroscopic and physical data with compounds reported in the literature. The structure of lobechine (1) was comprehensively established with the aid of 1D and 2D NMR spectroscopic analyses. In addition, selected isolates were screened for their inhibition of HSV-1 replication, superoxide anion generation, and elastase release. Among the tested compounds, scoparone (10) exhibited significant inhibition of superoxide anion generation with IC(50) of 6.14 ± 1.97 μM and lobechine (1) exhibited moderate inhibition of elastase release with IC(50) of 25.01 ± 6.95 μM, respectively.
Sigma-Aldrich
6,7-Dimethoxycoumarin, 98%
Pricing and availability is not currently available.