MilliporeSigma
  • Spectrophotometric determination of cobalt(II) and cyanocobalamin with vanillilfluorone and its applications.
Spectrophotometric determination of cobalt(II) and cyanocobalamin with vanillilfluorone and its applications.
Chemical & pharmaceutical bulletin (2011-06-02)
Mitsuru Hoshino, Momoka Matsushita, Megumi Samma, Mamiko Asano, Takako Yamaguchi, Yoshikazu Fujita
ABSTRACT
Spectrophotometric determination of cobalt(II) was accomplished with vanillilfluorone (VF) in the presence of dimethylbenzyltetradecylammonium chloride (Zephiramine, Zep). In the determination of cobalt(II), Beer's law was obeyed in the range of 24-470 ng/ml, with an effective molar absorption coefficient (at 575 nm) and relative standard deviation of 1.35×10(5) l mol(-1) cm(-1) and 0.66% (n=5), respectively. The composition ratio of the colored complex was determined by the mole ratio and continuous variation methods, and it was found to be Co(II) : VF : Zep=1 : 2 : 4. Analysis of cyanocobalamin by the same procedure showed that cyanocobalamin could be determined in the concentration range of 0.5-0.11 µg/ml using the proposed method.