MilliporeSigma
  • Alternative methods for testing treatment effects on the basis of multiple outcomes: simulation and case study.
Alternative methods for testing treatment effects on the basis of multiple outcomes: simulation and case study.
Statistics in medicine (2011-05-04)
Frank B Yoon, Garrett M Fitzmaurice, Stuart R Lipsitz, Nicholas J Horton, Nan M Laird, Sharon-Lise T Normand
ABSTRACT
In clinical trials multiple outcomes are often used to assess treatment interventions. This paper presents an evaluation of likelihood-based methods for jointly testing treatment effects in clinical trials with multiple continuous outcomes. Specifically, we compare the power of joint tests of treatment effects obtained from joint models for the multiple outcomes with univariate tests based on modeling the outcomes separately. We also consider the power and bias of tests when data are missing, a common feature of many trials, especially in psychiatry. Our results suggest that joint tests capitalize on the correlation of multiple outcomes and are more powerful than standard univariate methods, especially when outcomes are missing completely at random. When outcomes are missing at random, test procedures based on correctly specified joint models are unbiased, while standard univariate procedures are not. Results of a simulation study are reported, and the methods are illustrated in an example from the Clinical Antipsychotic Trials of Intervention Effectiveness for schizophrenia.
Sigma-Aldrich
Perphenazine
Pricing and availability is not currently available.