MilliporeSigma
  • Exceptionally fast uptake and metabolism of cyanidin 3-glucoside by rat kidneys and liver.
Exceptionally fast uptake and metabolism of cyanidin 3-glucoside by rat kidneys and liver.
Journal of natural products (2011-04-23)
Andreja Vanzo, Urska Vrhovsek, Federica Tramer, Fulvio Mattivi, Sabina Passamonti
ABSTRACT
To asses the hypothesis that anthocyanins are rapidly taken up from the blood into tissues, where they accumulate up to their bioactivity threshold, an intravenous dose of cyanidin 3-glucoside (1) was administered to anaesthetized rats. Cyanidin 3-glucoside (1) and its metabolites were analyzed in the plasma, kidneys, liver, urine, and bile, using last-generation mass spectrometry. Compound 1 was found to rapidly disappear from plasma (t/2=0.36 min). As soon as 15 s after its administration, both 1 and its methylation product, peonidin 3-glucoside (2), were detected in the plasma, kidneys, and liver. At 1 min, both 1 and 2 had almost disappeared from the plasma, but attained their peak concentrations in the kidneys and in the liver. Compound 2 was rapidly excreted both in the bile and in the urine. Three additional methylated metabolites were detected in traces, namely, delphinidin 3-glucoside (3), petunidin 3-glucoside (4), and malvidin 3-glucoside (5). These data contribute to solving the paradox of the high bioactivity of anthocyanins in spite of their apparent low bioavailability.
Sigma-Aldrich
Oenin chloride, ≥90% (HPLC)