MilliporeSigma
  • A rapid and sensitive liquid chromatography-tandem mass spectrometry method for determination of olopatadine concentration in human plasma.
A rapid and sensitive liquid chromatography-tandem mass spectrometry method for determination of olopatadine concentration in human plasma.
Journal of analytical toxicology (2011-03-15)
Ping Zhu, Yu-Guan Wen, Xiao-Ping Fan, Zhi-Ling Zhou, Rui-Xin Fan, Ji-Mei Chen, Ke-Li Huang, Xiao-Lan Zhu, Yan-Fang Chen, Jian Zhuang
ABSTRACT
A sensitive and rapid method based on liquid chromatography- tandem mass spectrometry (MS-MS) was developed for the determination of olopatadine in human plasma. Sample preparations were carried out by protein precipitation with the addition of acetonitrile followed by liquid-liquid extraction with ethyl acetate/dichloromethane after internal standard (IS, amitriptyline) spiked. After evaporation to dryness, the resultant residue was reconstituted in mobile phase. Separation of olopatadine and IS from the interferences was achieved on a C(18) column followed by MS-MS detection. The analytes were monitored in the positive ionization mode with a TurboIonspray source. Multiple reaction monitoring using the transition of m/z 338 → 165 and m/z 278 → 91 was performed to quantify olopatadine and IS, respectively. The method had a total chromatographic run time of 3.5 min and linear calibration curves over the concentration range of 0.2-100 ng/mL. The lower limit of quantification was 0.2 ng/mL. For each QC concentration level the intra- and interday precisions were less than 11.4%, and relative errors ranged between -6.40% and 9.26%. The validated method was successfully applied to the quantification of olopatadine concentration in human plasma after administration of olopatadine at an oral dose of 5 mg in order to evaluate the pharmacokinetics.
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)