MilliporeSigma
  • Rapid and sensitive liquid chromatography/tandem mass spectrometry method for simultaneous determination of enalapril and its major metabolite enalaprilat, in human plasma: application to a bioequivalence study.
Rapid and sensitive liquid chromatography/tandem mass spectrometry method for simultaneous determination of enalapril and its major metabolite enalaprilat, in human plasma: application to a bioequivalence study.
Drug testing and analysis (2011-02-23)
Chinmoy Ghosh, Ina Jain, Chandrakant P Shinde, Bhaswat S Chakraborty
ABSTRACT
A rapid and most sensitive method for simultaneous determination of enalapril (ENP) and its metabolite, enalaprilat (ENPT), in human plasma using ESI-LC-MS/MS (electrospray ionization liquid chromatography tandem mass spectrometry) positive ion multiple reactions monitoring (MRM) mode, was developed and validated. The procedure involves a simple solid-phase extraction (SPE) followed by evaporation of the sample. Chromatographic separation was carried out on a Hypurity C(18) column (50 mm × 4.6 mm, 5 µm) with an isocratic mobile phase and a total run time of 2.0 min only. The MRM of ENP and ENPT is 377.10 → 234.20 and 349.20 → 206.10 respectively. The standard calibration curves showed excellent linearity within the range of 0.064 to 431.806 ng/mL for ENA and 0.064 to 431.720 ng/mL for ENPT (r ≥ 0.990). This is the only method which can quantitate upto 0.064 ng/mL for both ENP and ENPT in a single run with the shortest analysis time. In matrix effect experiment, this method shows a % CV (% coefficients of variation) of less than 5, which means that the proposed method is free from any kind of irregular ionization process. This method was successfully applied to a pharmacokinetic study after oral administration of enalapril maleate 20 mg tablet in Indian healthy male volunteers.
Sigma-Aldrich
Enalaprilat dihydrate, ≥98% (HPLC)