MilliporeSigma
  • Ergodicity and efficiency of cross-polarization in NMR of static solids.
Ergodicity and efficiency of cross-polarization in NMR of static solids.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2011-02-08)
Alexander A Nevzorov
ABSTRACT
Cross-polarization transfer is employed in virtually every solid-state NMR experiment to enhance magnetization of low-gamma spins. Theory and experiment is used to assess the magnitude of the final quasistationary magnetization amplitude. The many-body density matrix equation is solved for relatively large (up to N=14) spin systems without the spin-temperature assumption for the final spin states. Simulations show that about 13% of the thermodynamic limit is still retained within the proton bath. To test this theoretical prediction, a combination of a reverse cross-polarization experiment and multiple contacts is employed to show that the thermodynamic limit of magnetization cannot be transferred from high- to low-gamma nuclei in a single contact. Multiple contacts, however, fully transfer the maximum magnetization. A simple diffusion on a cone model shows that slow dynamics can affect the build up profile for the transferred magnetization.
Sigma-Aldrich
N-Acetyl-L-leucine, ReagentPlus®, 99%
Pricing and availability is not currently available.