Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]yttrium
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur™, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium hydrogen difluoride, SAJ special grade, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Pricing and availability is not currently available.