MilliporeSigma
  • Synthesis and biological evaluation of novel N,N'-bis-methylenedioxybenzyl-alkylenediamines as bivalent anti-Alzheimer disease ligands.
Synthesis and biological evaluation of novel N,N'-bis-methylenedioxybenzyl-alkylenediamines as bivalent anti-Alzheimer disease ligands.
Journal of enzyme inhibition and medicinal chemistry (2011-01-22)
Wen Luo, Yan-Ping Li, Jia-Heng Tan, Lian-Quan Gu, Zhi-Shu Huang
ABSTRACT
A novel series of N,N'-bis-methylenedioxybenzyl-alkylenediamines 5a-5g have been designed, synthesized and evaluated as bivalent anti-Alzheimer's disease ligands. The enzyme inhibition assay results indicated that compounds 5e-5g inhibit both acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) in the micromolar range (IC(50), 2.76-4.24 µM for AChE and 3.02-5.14 µM for BuChE), which was in the same potential as the reference compound rivastigmine (IC(50), 5.50 µM for AChE and 1.60 µM for BuChE). It was found that compounds could bind simultaneously to the peripheral and catalytic sites of AChE. β-Amyloid (Aβ) aggregation inhibition assay results showed that compound 5e exhibited highest self-mediated Aβ fibril aggregation inhibition activity (40.3%) with a similar potential as curcumin (41.6%). It was also found that 5e-5g did not affect neuroblastoma cell viability at the concentration of 50 μM.
Sigma-Aldrich
2,2′-Dihydroxy-4-methoxybenzophenone, 98%
Pricing and availability is not currently available.
Supelco
Dioxybenzone, analytical standard
Pricing and availability is not currently available.