MilliporeSigma
  • Theoretical studies of molecular structure, electronic structure, spectroscopic properties and the ancillary ligand effect: a comparison of tris-chelate ML3-type and ML2X-type species for gallium(III) complexes with N,O-donor phenolic ligand, 2-(2-hydroxyphenyl)benzothiazole.
Theoretical studies of molecular structure, electronic structure, spectroscopic properties and the ancillary ligand effect: a comparison of tris-chelate ML3-type and ML2X-type species for gallium(III) complexes with N,O-donor phenolic ligand, 2-(2-hydroxyphenyl)benzothiazole.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-01-11)
Yi-Ping Tong, Yan-Wen Lin
ABSTRACT
Two Ga(III) complexes with main ligand, 2-(2-hydroxyphenyl)benzothiazole (HL'), namely mixed-ligand ML2X-type [GaL'2X'] (1) (HX'=acetic acid, as ancillary ligand) and the meridianal tris-chelate [GaL'3] (2) have been investigated by the density functional theory (DFT/TDDFT) level calculations. Both 1 and 2 can be presented as a similar "mixed-ligand ML2X-type" species. The molecular geometries, electronic structures, metal-ligand bonding property of Ga-O (N) (main ligand), Ga-O (N) (ancillary ligand) interactions, and the ancillary ligand effect on their HOMO-LUMO gap, their absorption/emission property, and their absorption/emission wavelengths/colors for them have been discussed in detail based on the orbital interactions, the partial density of states (PDOS), and so on. The current investigation also indicates that it is quite probable that by introduction of different ancillary ligands, a series of new mixed-ligand ML2X-type complexes for group 13 metals can be designed with their absorption/emission property and the absorption/emission wavelengths and colors being tuned.
Sigma-Aldrich
2-(2-Hydroxyphenyl)benzothiazole, 97%
Pricing and availability is not currently available.