MilliporeSigma
  • N-aminoazetidinecarboxylic acid: direct access to a small-ring hydrazino acid.
N-aminoazetidinecarboxylic acid: direct access to a small-ring hydrazino acid.
The Journal of organic chemistry (2010-12-25)
Valérie Declerck, David J Aitken
ABSTRACT
A short and efficient synthesis of the previously unknown N-aminoazetidinecarboxylic acid has been established using a photochemical [2 + 2] cycloaddition strategy starting from 6-azauracil. Chiral derivatization with a nonracemic oxazolidinone provided access to both enantiomers of the title product.
Sigma-Aldrich
6-Azauracil, ≥98%