MilliporeSigma
  • The occurrence of the herbicide dalapon (2,2-dichloropropionate) in potable water as a disinfection by-product.
The occurrence of the herbicide dalapon (2,2-dichloropropionate) in potable water as a disinfection by-product.
Journal of environmental monitoring : JEM (2010-12-18)
Darryl W Hawker, Janet L Cumming, Andrew Watkinson, Michael E Bartkow
ABSTRACT
Salts of 2,2-dichloropropionic acid, such as dalapon, are well known as herbicides and are regulated as such in potable water in Australia and elsewhere. It is also an identified disinfection by-product (DBP), but little is known about the compound's formation and typical levels from this source. This work presents results from a sampling campaign where 2,2-dichloropropionate was found at levels between 0.1 and 0.5 µg l(-1) in potable water samples from a major treatment plant in South East Queensland, Australia. However, levels were below the reporting limit (0.01 µg l(-1)) in the immediate source water for the plant. Also, temporal trends in 2,2-dichloropropionate observed in treated water during sampling mirrored those of trihalomethanes albeit at much lower concentrations, suggesting that the occurrence is due to in situ formation as a DBP. This could present a regulatory dilemma in some jurisdictions.
Product Description
Supelco
Dalapon, PESTANAL®, analytical standard
Pricing and availability is not currently available.