MilliporeSigma
Inactivation of Ascaris suum by short-chain fatty acids.
Applied and environmental microbiology (2010-11-09)
Michael A Butkus, Kelly T Hughes, Dwight D Bowman, Janice L Liotta, Michael B Jenkins, Michael P Labare
ABSTRACT
Ascaris suum eggs were inactivated in distilled water and digested sludge by butanoic, pentanoic, and hexanoic acids. The fatty acids (short-chain fatty acids [SCFA]) were effective only when protonated and at sufficient concentrations. The conjugate bases were not effective at the concentrations evaluated. Predictions from an inhibition model (50% inhibitory concentration [IC(50)]) based on quantitative structure-activity relationships were congruent with inactivation data.
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium hexanoate, 99-100%
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Sigma-Aldrich
Hexanoic acid, ≥99%
Supelco
Hexanoic acid, analytical standard