MilliporeSigma
  • Modification of polydivinylbenzene microspheres by a hydrobromination/click-chemistry protocol and their protein-adsorption properties.
Modification of polydivinylbenzene microspheres by a hydrobromination/click-chemistry protocol and their protein-adsorption properties.
Macromolecular bioscience (2010-10-15)
Yasemin Yuksel Durmaz, Bunyamin Karagoz, Niyazi Bicak, Dilek Odaci Demirkol, Esra Evrim Yalcinkaya, Suna Timur, Yusuf Yagci
ABSTRACT
Hydrophobic- and/or hydrophilic-polymer-grafted PDVB microspheres are synthesized by the combination of hydrobromination and click-chemistry processes. The modified-PDVB microspheres and the intermediates at various stages of synthesis are characterized using GPC, ¹H NMR and FTIR spectroscopy and TGA analysis. Use of the microspheres as a support matrix for reversible protein immobilization via adsorption is investigated. The system parameters such as the adsorption conditions (i.e., enzyme concentration, medium pH) and desorption are studied and evaluated with regards to the biocatalytic activity and adsorption capacity.
Product Description
Supelco
Amberlite XAD16N, 20-60 mesh
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%