MilliporeSigma
  • Paraoxon, 4-nitrophenyl phosphate and acetate are substrates of α- but not of β-, γ- and ζ-carbonic anhydrases.
Paraoxon, 4-nitrophenyl phosphate and acetate are substrates of α- but not of β-, γ- and ζ-carbonic anhydrases.
Bioorganic & medicinal chemistry letters (2010-09-14)
Alessio Innocenti, Claudiu T Supuran
ABSTRACT
Carbonic anhydrases (CAs, EC 4.2.1.1) belonging to α-, β-, γ- and ζ-classes and from various organisms, ranging from the bacteria, archaea to eukarya domains, were investigated for their esterase/phosphatase activity with 4-nitrophenyl acetate, 4-nitrophenyl phosphate and paraoxon as substrates. Only α-CAs showed esterase/phosphatase activity, whereas enzymes belonging to the β-, γ- and ζ-classes were completely devoid of such activity. Paraoxon, the metabolite of the organophosphorus insecticide parathione, was a much better substrate for several human/murine α-CA isoforms (CA I, II and XIII), with k(cat)/K(M) in the range of 2681.6-4474.9M(-1)s(-1), compared to 4-nitrophenyl phosphate (k(cat)/K(M) of 14.9-1374.4M(-1)s(-1)).
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate