MilliporeSigma
  • Pharmacokinetic and tissue distribution of doxycycline in broiler chickens pretreated with either: diclazuril or halofuginone.
Pharmacokinetic and tissue distribution of doxycycline in broiler chickens pretreated with either: diclazuril or halofuginone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-08-26)
A Y I El-Gendi, M Atef, Aziza M M Amer, Gehan M Kamel
ABSTRACT
Following IV injection of doxycycline in a dose of 20 mg kg(-1) b.wt., its serum concentration was best fitted in two-compartment open model in chickens fed either on control or on anticoccidials-containing rations. Diclazuril and halofuginone resulted in a significant short distribution half-life (t(½α)) (7.17±0.39 and 11.88±1.05 min, respectively) and increased total body clearance (Cl(tot)) 0.37±0.024 and 0.295±0.034 L/kg/h, respectively. Following oral dosing the tested drug absorbed with t(½ab) of 41.38±1.6, 17.48±0.86 and 41.83±1.8 min, respectively and their C(max) values (3.18±0.18, 5.425±0.48 and 0.986±0.037 μg/ml) were attained at 2.07±0.097, 1.403±0.074 and 2.55±0.106 h. For doxycycline alone and in presence of diclazuril and halofuginone, respectively. Systemic bioavailability was 22.64±3.46, 86.74±9.23 and 22.38±3.09%, respectively. Following IM injection t(½ab) were 9.096±1.34 for doxycycline alone, 16.24±2.21 and 15.6±1.7 min in the presence of diclazuril and halofuginone, respectively. C(max) was 3.10±0.28, 4.63±0.57 and 0.55±0.07 μg/ml reached at 0.8±0.083, 1.13±0.126 and 1.21±0.105 h. For the antibiotic alone, and in presence of either diclazuril and halofuginone, respectively. Systemic bioavailability was 22.41±3.86, 88.97±12.9 and 12.31±0.99% in chickens fed on anticoccidial-free, diclazuril- and halofuginone-containing rations, respectively. Both the tested anticoccidials induced higher doxycycline tissue residues in all tested tissue samples.
Product Description
Diclazuril for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Diclazuril, VETRANAL®, analytical standard
Pricing and availability is not currently available.