MilliporeSigma
  • Absorption spectroscopy of adenine, 9-methyladenine, and 2-aminopurine in helium nanodroplets.
Absorption spectroscopy of adenine, 9-methyladenine, and 2-aminopurine in helium nanodroplets.
Physical chemistry chemical physics : PCCP (2010-08-03)
Szymon Smolarek, Anouk M Rijs, Wybren Jan Buma, Marcel Drabbels
ABSTRACT
High-resolution absorption spectra of adenine, 9-methyladenine and 2-aminopurine in helium nanodroplets have been recorded. In contrast to molecular beam experiments, large variations in linewidths are observed for adenine and 9-methyladenine. At the same time, the spectrum of 2-aminopurine remains sharp upon solvation in helium droplets. The line broadening observed for adenine and 9-methyladenine is attributed to a significant decrease of the lifetime of the (1)L(b)(ππ*) state and of (1)nπ* levels vibronically coupled to this state. The origin of the lifetime reduction is argued to be related to the increased accessibility of the (1)nπ*/(1)L(b)(ππ*) conical intersection upon solvation of these molecules in liquid helium.
Sigma-Aldrich
9-Methyladenine, 97%