MilliporeSigma
  • Inhibition of chorismate-utilising enzymes by 2-amino-4-carboxypyridine and 4-carboxypyridone and 5-carboxypyridone analogues.
Inhibition of chorismate-utilising enzymes by 2-amino-4-carboxypyridine and 4-carboxypyridone and 5-carboxypyridone analogues.
Organic & biomolecular chemistry (2010-06-10)
Richard J Payne, Esther M M Bulloch, Olivier Kerbarh, Chris Abell
ABSTRACT
Several 2-amino-4-carboxypyridine, 4- and 5-carboxypyridone-based compounds were prepared and tested against three members of the chorismate-utilising enzyme family, anthranilate synthase, isochorismate synthase and salicylate synthase. Most compounds exhibited low micromolar inhibition of these three enzymes. The most potent inhibitor was a 4-carboxypyridone analogue bearing a lactate side chain on the pyridyl nitrogen which exhibited inhibition constants of 5, 91 and 54 muM against anthranilate synthase, isochorismate synthase and salicylate synthase respectively.
Sigma-Aldrich
Chorismic acid from Enterobacter aerogenes, ≥80%