MilliporeSigma
  • Molecular study on copper-mediated tumor proteasome inhibition and cell death.
Molecular study on copper-mediated tumor proteasome inhibition and cell death.
International journal of oncology (2010-06-02)
Yan Xiao, D I Chen, Xia Zhang, Qiuzhi Cui, Yuhua Fan, Caifeng Bi, Q Ping Dou
ABSTRACT
The metal ion copper is a cofactor essential for maintaining normal biological and physical functions in human beings. High copper levels have been found in variety of tumor tissues and are involved in tumor angiogenesis processes. The ubiquitin-proteasome system plays an important role in cell growth and apoptosis and has been shown as a novel target for cancer therapy. We previously reported that some organic copper complexes can inhibit the proteasomal chymotrypsin-like activity and induce apoptosis in human cancer cells and xenograft models. In the current study, we investigated the effect of oxidation status of copper, Cu(I) or Cu(II), on inhibition of proteasome activity, induction of apoptosis, and induction of reactive oxygen species (ROS) in human cancer cells. We report four major findings here: i) both Cu(I) and Cu(II) could inhibit the chymotrypsin-like activity of purified 20S proteasome, but Cu(I) was more potent than Cu(II), ii) purified 20S proteasome protein was able to reduce Cu(II) to Cu(I), suggesting that Cu(I) is the oxidation status of copper that directly reacts with the proteasome, iii) when complexed with the copper ligand neocuproine, Cu(I) showed higher ability to induce ROS production in cancer cells, compared with Cu(II), iv) addition of a ROS scavenger in the cancer cell culture-blocked copper-induced ROS generation, but did not overcome copper-mediated proteasome-inhibitory and cell death-inducing events, demonstrating the ROS-independent proteasome-inhibitory property of copper complexes.
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(I) chloride, JIS special grade, ≥95.0%
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Neocuproine, ≥98%