MilliporeSigma
  • Synthesis of (-)-paroxetine via enantioselective phase-transfer catalytic monoalkylation of malonamide ester.
Synthesis of (-)-paroxetine via enantioselective phase-transfer catalytic monoalkylation of malonamide ester.
Organic letters (2010-05-27)
Mi-Hyun Kim, Yohan Park, Byeong-Seon Jeong, Hyeung-Geun Park, Sang-Sup Jew
ABSTRACT
A new enantioselective synthetic method of (-)-paroxetine is reported. (-)-Paroxetine could be obtained in 15 steps (95% ee and 9.1% overall yield) from N,N-bis(p-methoxyphenyl)malonamide tert-butyl ester via the enantioselective phase-transfer catalytic alkylation and the diastereoselective Michael addition as the key steps.
Sigma-Aldrich
Malonamide, 97%