MilliporeSigma
  • Attenuation of oxidative stress and alteration of hepatic tissue ultrastructure by D-pinitol in streptozotocin-induced diabetic rats.
Attenuation of oxidative stress and alteration of hepatic tissue ultrastructure by D-pinitol in streptozotocin-induced diabetic rats.
Free radical research (2010-04-08)
Selvaraj Sivakumar, Periyasamy Palsamy, Sorimuthu Pillai Subramanian
ABSTRACT
The present study was aimed to investigate the effect of D-pinitol on hyperglycaemia mediated oxidative stress by analysing the hepatic antioxidant competence, pro-inflammatory cytokines and ultrastructural changes in liver tissues of streptozotocin-induced diabetic rats. Oral administration of D-pinitol (50 mg/kg b.w.) resulted in significant (p < 0.05) attenuation in blood glucose, glycosylated haemoglobin and pro-inflammatory markers such as TNF-alpha, IL-1beta, IL-6, NF-kappaB p65 unit and NO and significant (p < 0.05) elevation in the plasma insulin level. In addition, D-pinitol instigated a significant escalation in the levels of hepatic tissue non-enzymatic antioxidants and the activities enzymatic antioxidants of diabetic rats with significant (p < 0.05) decrease in lipid peroxides and hydroperoxides formation, thus demonstrating the protective role of D-pinitol on the hepatic tissues from oxidative stress-induced liver damage. These biochemical observations were complemented by histological and ultrastructural examination of liver section. Thus, the present study demonstrates the hepatoprotective nature of D-pinitol by attenuating hyperglycaemia-mediated pro-inflammatory cytokines and oxidative stress.
Sigma-Aldrich
D-Pinitol, 95%