MilliporeSigma
  • NK cells influence both innate and adaptive immune responses after mucosal immunization with antigen and mucosal adjuvant.
NK cells influence both innate and adaptive immune responses after mucosal immunization with antigen and mucosal adjuvant.
Journal of immunology (Baltimore, Md. : 1950) (2010-03-12)
Lindsay J Hall, Simon Clare, Gordon Dougan
ABSTRACT
NK cells were found to be recruited in a temporally controlled manner to the nasal-associated lymphoid tissue and the cervical lymph nodes of mice after intranasal immunization with Ag85B-early secreted antigenic target 6 kDa from Mycobacterium tuberculosis mixed with Escherichia coli heat-labile toxin as adjuvant. These NK cells were activated and secreted a diverse range of cytokines and other immunomodulators. Using Ab depletion targeting anti-asialo GM1, we found evidence for altered trafficking, impaired activation, and cytokine secretion of dendritic cells, macrophages, and neutrophils in immunized NK cell-depleted mice compared with control animals. Analysis of Ag-specific immune responses revealed an attenuated Ab and cytokine response in immunized NK cell-depleted animals. Systemic administration of rIL-6 but not rIFN-gamma significantly restored immune responses in mice depleted of NK cells. In conclusion, cytokine production, particularly IL-6, via NK cells and NK cell-activated immune populations plays an important role in the establishment of local innate immune responses and the consequent development of adaptive immunity after mucosal immunization.
Sigma-Aldrich
Asialoganglioside GM1 from bovine brain, ~98%, lyophilized powder