MilliporeSigma
  • 3-hydroxyphthalic anhydride-modified chicken ovalbumin exhibits potent and broad anti-HIV-1 activity: a potential microbicide for preventing sexual transmission of HIV-1.
3-hydroxyphthalic anhydride-modified chicken ovalbumin exhibits potent and broad anti-HIV-1 activity: a potential microbicide for preventing sexual transmission of HIV-1.
Antimicrobial agents and chemotherapy (2010-03-03)
Lin Li, Lili He, Suiyi Tan, Xinhua Guo, Hong Lu, Zhi Qi, Chungen Pan, Xiuli An, Shibo Jiang, Shuwen Liu
ABSTRACT
Heterosexual transmission is the primary route by which women acquire human immunodeficiency virus (HIV)/AIDS. Thus, development of woman-controlled topical microbicides for prevention of sexual transmission of HIV is urgently needed. Here we report that 3-hydroxyphthalic anhydride-modified chicken ovalbumin (HP-OVA) exhibits potent antiviral activity against a broad spectrum of human immunodeficiency virus type 1 (HIV-1) isolates with different genotypes and biotypes. Its antiviral activity is correlated with the percentages of the chemically modified and unmodified lysines and arginines in OVA. HP-OVA inhibits HIV-1 fusion and entry through multiple mechanisms of action, including (i) blocking gp120 binding to CD4 and (ii) interfering with gp41 six-helix bundle formation. Because of the widespread availability and established safety profile of OVA, HP-OVA has good potential to be developed as an effective, safe, and affordable microbicide for prevention of HIV sexual transmission.
Sigma-Aldrich
3-Hydroxyphthalic anhydride, 98%